Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMTOR3
Entrez Gene ID
709133
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 3
Gene Aliases
EGK_15962, MAPKSP1
Chromosome Location
Chr.5: 98243578 - 98257692 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257535.1 NP_001244464.1 - 462 98
GenBank mRNA JU320556.1 - - 462 98
JU471004.1 - - 426 98
JV044392.1 - - 442 98

View Full Product Details