Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPX4
Entrez Gene ID
705333
Gene Name
glutathione peroxidase 4
Gene Aliases
-
Chromosome Location
Chr.19: 879102 - 881951 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001118889.2 NP_001112361.1 - 398 69
NM_001346420.1 NP_001333349.1 - 526 69
GenBank mRNA CB552751.1 - - 103 69
CN643832.1 - - 299 69
JV046824.1 - - 332 69
JV465885.1 - - 329 69
JV602446.1 - - 526 69
JV649198.1 - - 397 69
JV667383.1 - - 452 69
JV681449.1 - - 384 69

View Full Product Details