Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THAP11
Entrez Gene ID
700354
Gene Name
THAP domain containing 11
Gene Aliases
-
Chromosome Location
Chr.20: 55056866 - 55058958 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015126436.2 XP_014981922.1 - 1797 97
GenBank mRNA JU325697.1 - - 1049 97
JU325698.1 - - 1044 97
JV719343.1 - - 1078 97

View Full Product Details