Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DMRT3
Entrez Gene ID
697323
Gene Name
doublesex and mab-3 related transcription factor 3
Gene Aliases
-
Chromosome Location
Chr.15: 82855282 - 82871114 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001089961.4 XP_001089961.2 - 867 105

View Full Product Details