Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELK4
Entrez Gene ID
699281
Gene Name
ETS transcription factor ELK4
Gene Aliases
EGK_01741
Chromosome Location
Chr.1: 63130130 - 63154206 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261604.1 NP_001248533.1 - 412 128
XM_015118422.2 XP_014973908.1 - 299 128
GenBank mRNA JU333301.1 - - 356 128
JU474615.1 - - 412 128

View Full Product Details