Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC22
Entrez Gene ID
710205
Gene Name
DnaJ heat shock protein family (Hsp40) member C22
Gene Aliases
-
Chromosome Location
Chr.11: 48957311 - 48962713 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194448.2 NP_001181377.1 - 292 115

View Full Product Details