Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WFIKKN1
Entrez Gene ID
722338
Gene Name
WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.20: 657028 - 661998 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JV565063.1 - - 1572 112
JV641764.1 - - 1582 112

View Full Product Details