Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RLN3
Entrez Gene ID
717577
Gene Name
relaxin 3
Gene Aliases
RLX3
Chromosome Location
Chr.19: 13611425 - 13636120 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104576.1 NP_001098046.1 - 195 144
XM_015123081.2 XP_014978567.1 - 1113 144
GenBank mRNA DQ288862.1 - - 195 144

View Full Product Details