Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TGIF2LX
Entrez Gene ID
574239
Gene Name
TGFB induced factor homeobox 2 like X-linked
Gene Aliases
TGIFLX
Chromosome Location
Chr.X: 86732685 - 86734106 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032888.1 NP_001028060.1 - 794 181
XM_015127766.2 XP_014983252.2 - 772 181
GenBank mRNA AJ345078.2 - - 794 181

View Full Product Details