Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRGPRF
Entrez Gene ID
721546
Gene Name
MAS related GPR family member F
Gene Aliases
MGRC
Chromosome Location
Chr.14: 5783478 - 5793545 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015113470.2 XP_014968956.2 - 666 64
XM_015113471.2 XP_014968957.2 - 453 64
XM_028833100.1 XP_028688933.1 - 265 64
XM_028833101.1 XP_028688934.1 - 381 64
XM_028833102.1 XP_028688935.1 - 628 64
XM_028833103.1 XP_028688936.1 - 368 64
XM_028833104.1 XP_028688937.1 - 268 64
GenBank mRNA JV044488.1 - - 237 64
JV635835.1 - - 227 64

View Full Product Details