Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKX2-3
Entrez Gene ID
708540
Gene Name
NK2 homeobox 3
Gene Aliases
-
Chromosome Location
Chr.9: 100004550 - 100009116 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001106216.4 XP_001106216.2 - 1069 148

View Full Product Details