Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRFAP1L1
Entrez Gene ID
710411
Gene Name
Morf4 family associated protein 1 like 1
Gene Aliases
EGK_15549
Chromosome Location
Chr.5: 6260630 - 6262865 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193928.1 NP_001180857.1 - 1188 129

View Full Product Details