Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANKRD9
Entrez Gene ID
693496
Gene Name
ankyrin repeat domain 9
Gene Aliases
-
Chromosome Location
Chr.7: 164750003 - 164752827 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015144527.2 XP_015000013.2 - 97 84
XM_015144528.2 XP_015000014.2 - 89 84

Gene Symbol
LOC114679686
Entrez Gene ID
114679686
Gene Name
uncharacterized LOC114679686
Gene Aliases
-
Chromosome Location
Chr.7: 164751189 - 164754704 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028851995.1 XP_028707828.1 - 557 84

View Full Product Details