Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMUB2
Entrez Gene ID
714541
Gene Name
transmembrane and ubiquitin like domain containing 2
Gene Aliases
EGK_08797
Chromosome Location
Chr.16: 55449292 - 55454628 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194640.2 NP_001181569.1 - 988 124
XM_015119759.2 XP_014975245.1 - 933 124
XM_015119761.2 XP_014975247.1 - 849 124
XM_015119762.2 XP_014975248.1 - 823 124
XM_015119763.2 XP_014975249.1 - 747 124
XM_028835663.1 XP_028691496.1 - 1273 124
XM_028835664.1 XP_028691497.1 - 795 124
XM_028835666.1 XP_028691499.1 - 873 124
GenBank mRNA JU322510.1 - - 863 124
JU322512.1 - - 692 124
JU477171.1 - - 1077 124
JV048224.1 - - 1077 124
JV566196.1 - - 1110 124

View Full Product Details