Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC704445
Entrez Gene ID
704445
Gene Name
keratin-associated protein 13-3
Gene Aliases
EGK_13325, KRTAP13-3
Chromosome Location
Chr.3: 16217486 - 16218219 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028845174.1 XP_028701007.1 - 271 143

View Full Product Details