Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP7A1
Entrez Gene ID
698127
Gene Name
cytochrome P450, family 7, subfamily A, polypeptide 1
Gene Aliases
-
Chromosome Location
Chr.8: 59141315 - 59154866 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193803.3 NP_001180732.3 - 461 177
XM_015145392.2 XP_015000878.2 - 428 177

View Full Product Details