Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRA2
Entrez Gene ID
703894
Gene Name
gamma-aminobutyric acid type A receptor subunit alpha2
Gene Aliases
EGK_15690
Chromosome Location
Chr.5: 45572754 - 45726589 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261668.1 NP_001248597.1 - 1191 139
XM_015138315.2 XP_014993801.1 - 1334 139
XM_015138316.2 XP_014993802.1 - 1402 139
XM_028848079.1 XP_028703912.1 - 1112 139
XM_028848080.1 XP_028703913.1 - 1401 139
GenBank mRNA JU333815.1 - - 1167 139
JU333816.1 - - 1132 139
JU333817.1 - - 1191 139
JV711309.1 - - 1141 139

View Full Product Details