Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OR4D5
Entrez Gene ID
709394
Gene Name
olfactory receptor, family 4, subfamily D, member 5
Gene Aliases
-
Chromosome Location
Chr.14: 116850579 - 116851673 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194427.2 NP_001181356.2 - 809 128

View Full Product Details