Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DBX2
Entrez Gene ID
701170
Gene Name
developing brain homeobox 2
Gene Aliases
-
Chromosome Location
Chr.11: 44468703 - 44512381 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001092529.4 XP_001092529.2 - 780 148
GenBank mRNA JU325749.1 - - 111 148

View Full Product Details