Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HMCES
Entrez Gene ID
703524
Gene Name
5-hydroxymethylcytosine binding, ES cell specific
Gene Aliases
C11H3orf37, C3orf37
Chromosome Location
Chr.2: 147197406 - 147227337 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194040.1 NP_001180969.1 - 580 96
XM_001095958.4 XP_001095958.1 - 537 96
XM_015130134.2 XP_014985620.1 - 498 96
XM_028843211.1 XP_028699044.1 - 668 96
XM_028843212.1 XP_028699045.1 - 490 96
GenBank mRNA JU335538.1 - - 407 96
JU476466.1 - - 410 96
JV513579.1 - - 364 96
JV587837.1 - - 406 96
JV656419.1 - - 311 96

View Full Product Details