Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IP6K1
Entrez Gene ID
705576
Gene Name
inositol hexakisphosphate kinase 1
Gene Aliases
EGK_11628
Chromosome Location
Chr.2: 105840252 - 105912723 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257763.1 NP_001244692.1 - 1102 100
XM_015130794.2 XP_014986280.1 - 1496 100
XM_015130795.2 XP_014986281.1 - 937 100
XM_015130796.2 XP_014986282.1 - 940 100
XM_028843239.1 XP_028699072.1 - 936 100
GenBank mRNA JU337334.1 - - 1102 100
JU478036.1 - - 1098 100
JV043613.1 - - 1108 100
JV666759.1 - - 1110 100

View Full Product Details