Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB37
Entrez Gene ID
698842
Gene Name
RAB37, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.16: 71007508 - 71084905 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266608.1 NP_001253537.1 - 138 103
GenBank mRNA JU475398.1 - - 138 103

View Full Product Details