Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GOSR2
Entrez Gene ID
716841
Gene Name
golgi SNAP receptor complex member 2
Gene Aliases
-
Chromosome Location
Chr.16: 57038338 - 57066256 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257778.1 NP_001244707.1 - 95 130
XM_015119780.2 XP_014975266.1 - 130 130
XM_028835739.1 XP_028691572.1 - 130 130
XM_028835740.1 XP_028691573.1 - 130 130
XM_028835741.1 XP_028691574.1 - 130 130
GenBank mRNA JU332253.1 - - 95 130
JU472931.1 - - 98 130
JV043835.1 - - 94 130
JV529778.1 - - 130

View Full Product Details