Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAV2
Entrez Gene ID
706980
Gene Name
caveolin 2
Gene Aliases
-
Chromosome Location
Chr.3: 142725100 - 142734248 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168619.1 NP_001162090.1 - 3174 150

View Full Product Details