Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SBK1
Entrez Gene ID
707411
Gene Name
SH3 domain binding kinase 1
Gene Aliases
-
Chromosome Location
Chr.20: 26609490 - 26676537 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015125847.2 XP_014981333.2 - 1109 85
XM_028841505.1 XP_028697338.1 - 1050 85
GenBank mRNA JV616219.1 - - 972 85

View Full Product Details