Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELP6
Entrez Gene ID
711423
Gene Name
elongator acetyltransferase complex subunit 6
Gene Aliases
C2H3orf75, C3orf75
Chromosome Location
Chr.2: 103512145 - 103530920 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001113386.4 XP_001113386.1 - 298 106
GenBank mRNA JU323417.1 - - 218 106
JU473382.1 - - 230 106
JV045576.1 - - 234 106
JV529782.1 - - 187 106

View Full Product Details