Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP28
Entrez Gene ID
574250
Gene Name
matrix metallopeptidase 28
Gene Aliases
-
Chromosome Location
Chr.16: 32112299 - 32154957 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278395.2 NP_001265324.1 - 242 90
NM_001278396.2 NP_001265325.1 - 242 90
XM_015119023.2 XP_014974509.1 - 435 90

View Full Product Details