Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TYW5
Entrez Gene ID
699197
Gene Name
tRNA-yW synthesizing protein 5
Gene Aliases
C2orf60, JMJD5
Chromosome Location
Chr.12: 87030462 - 87059360 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193883.2 NP_001180812.2 - 601 131
XM_015110595.2 XP_014966081.2 - 839 131
XM_015110596.2 XP_014966082.2 - 839 131
XM_015110597.2 XP_014966083.1 - 750 131
XM_028830441.1 XP_028686274.1 - 802 131
GenBank mRNA JU338818.1 - - 581 131

View Full Product Details