Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REC114
Entrez Gene ID
699749
Gene Name
REC114 meiotic recombination protein
Gene Aliases
-
Chromosome Location
Chr.7: 50275478 - 50394223 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001093604.4 XP_001093604.1 - 581 134
XM_015142681.2 XP_014998167.1 - 646 134

View Full Product Details