Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRD5A1
Entrez Gene ID
695103
Gene Name
steroid 5 alpha-reductase 1
Gene Aliases
-
Chromosome Location
Chr.6: 6552085 - 6589567 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001083405.4 XP_001083405.1 - 902 109
XM_028849331.1 XP_028705164.1 - 875 109
GenBank mRNA JV659654.1 - - 541 109

View Full Product Details