Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BMP3
Entrez Gene ID
695432
Gene Name
bone morphogenetic protein 3
Gene Aliases
-
Chromosome Location
Chr.5: 52641224 - 52666699 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001089525.4 XP_001089525.2 - 2011 138

View Full Product Details