Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EFNA5
Entrez Gene ID
704351
Gene Name
ephrin A5
Gene Aliases
EGK_16736
Chromosome Location
Chr.6: 104830770 - 105125099 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001265728.1 NP_001252657.1 - 376 143
XM_015140527.2 XP_014996013.1 - 852 143
GenBank mRNA JU321748.1 - - 154 143
JV565076.1 - - 376 143
JV708763.1 - - 353 143

View Full Product Details