Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- DMTN
- Entrez Gene ID
-
706762
- Gene Name
- dematin actin binding protein
- Gene Aliases
-
EPB49
- Chromosome Location
-
Chr.8: 22214344 - 22248806 on Build Mmul_10
View Full Product Details