Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFB5
Entrez Gene ID
708821
Gene Name
NADH:ubiquinone oxidoreductase subunit B5
Gene Aliases
EGK_11875
Chromosome Location
Chr.2: 84755189 - 84774032 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001266533.2 NP_001253462.1 - 366 96
XM_015131545.2 XP_014987031.2 - 243 96
GenBank mRNA JU325229.1 - - 298 96
JU473772.1 - - 351 96
JV636173.1 - - 368 96
JV715754.1 - - 320 96

View Full Product Details