Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC16A1
Entrez Gene ID
706593
Gene Name
solute carrier family 16 member 1
Gene Aliases
EGK_01106
Chromosome Location
Chr.1: 111271542 - 111314684 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001257704.1 NP_001244633.1 - 615 151
XM_015147143.2 XP_015002629.2 - 540 151
XM_015147153.2 XP_015002639.2 - 1196 151
GenBank mRNA JU323021.1 - - 615 151
JU473026.1 - - 664 151
JV721619.1 - - 623 151

View Full Product Details