Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KMT2D
Entrez Gene ID
710795
Gene Name
lysine methyltransferase 2D
Gene Aliases
MLL2
Chromosome Location
Chr.11: 48628136 - 48670741 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015151647.2 XP_015007133.2 - 6387 77

View Full Product Details