Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STX10
Entrez Gene ID
717208
Gene Name
syntaxin 10
Gene Aliases
EGK_10192
Chromosome Location
Chr.19: 12737719 - 12744310 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261693.1 NP_001248622.1 - 42 60
XM_015122994.1 XP_014978480.1 - 233 60
GenBank mRNA JU336890.1 - - 37 60
JU477631.1 - - 42 60

View Full Product Details