Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
URI1
Entrez Gene ID
700838
Gene Name
URI1 prefoldin like chaperone
Gene Aliases
C19H19orf2
Chromosome Location
Chr.19: 29372383 - 29464740 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001085459.4 XP_001085459.3 - 1769 169
XM_015123477.2 XP_014978963.2 - 1332 169
XM_015123478.2 XP_014978964.2 - 1422 169
XM_015123480.2 XP_014978966.2 - 1301 169
XM_015123483.2 XP_014978969.2 - 1732 169
XM_028839928.1 XP_028695761.1 - 1605 169
GenBank mRNA JV664417.1 - - 1441 169
JV699930.1 - - 1419 169

View Full Product Details