Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAP23
Entrez Gene ID
708225
Gene Name
synaptosome associated protein 23
Gene Aliases
EGK_17405
Chromosome Location
Chr.7: 19001556 - 19043337 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261338.1 NP_001248267.1 - 663 110
XM_015141895.2 XP_014997381.1 - 1069 110
XM_015141896.2 XP_014997382.1 - 707 110
XM_015141898.2 XP_014997384.1 - 638 110
XM_015141899.2 XP_014997385.1 - 642 110
XM_015141900.2 XP_014997386.1 - 671 110
XM_015141901.2 XP_014997387.1 - 877 110
GenBank mRNA JU336916.1 - - 652 110
JU336917.1 - - 652 110
JU477651.1 - - 665 110
JV048584.1 - - 663 110

View Full Product Details