Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIGLEC5
Entrez Gene ID
719730
Gene Name
sialic acid binding Ig like lectin 5
Gene Aliases
-
Chromosome Location
Chr.19: 51369186 - 51385563 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109886.1 NP_001103356.1 - 61 99
GenBank mRNA EU131368.1 - - 61 99
EU131371.1 - - 61 99

View Full Product Details