Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BUD31
- Entrez Gene ID
-
718694
- Gene Name
- BUD31 homolog
- Gene Aliases
-
EGK_13487
- Chromosome Location
-
Chr.3: 41522847 - 41533027 on Build Mmul_10
- Gene Symbol
- LOC718712
- Entrez Gene ID
-
718712
- Gene Name
- pentatricopeptide repeat-containing protein 1, mitochondrial
- Gene Aliases
-
EGK_13488, PTCD1
- Chromosome Location
-
Chr.3: 41532532 - 41550171 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JU484148.1 |
- |
- |
2977 |
150 |
JV647815.1 |
- |
- |
2972 |
150 |
View Full Product Details