Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELAVL2
Entrez Gene ID
708195
Gene Name
ELAV like RNA binding protein 2
Gene Aliases
-
Chromosome Location
Chr.15: 59867378 - 60026998 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261000.1 NP_001247929.1 - 730 91
XM_015117598.2 XP_014973084.1 - 1134 91
XM_015117599.2 XP_014973085.1 - 615 91
XM_015117600.2 XP_014973086.1 - 1134 91
XM_015117601.2 XP_014973087.1 - 1131 91
XM_015117602.2 XP_014973088.1 - 1134 91
XM_015117603.2 XP_014973089.1 - 1134 91
XM_015117604.2 XP_014973090.1 - 930 91
XM_015117605.2 XP_014973091.1 - 750 91
XM_015117606.2 XP_014973092.1 - 1043 91
XM_015117607.2 XP_014973093.1 - 1043 91
XM_028834315.1 XP_028690148.1 - 791 91
XM_028834316.1 XP_028690149.1 - 841 91
XM_028834317.1 XP_028690150.1 - 1131 91
XM_028834318.1 XP_028690151.1 - 841 91
XM_028834319.1 XP_028690152.1 - 722 91
XM_028834320.1 XP_028690153.1 - 724 91
XM_028834321.1 XP_028690154.1 - 634 91
XM_028834322.1 XP_028690155.1 - 521 91
XM_028834324.1 XP_028690157.1 - 697 91
XM_028834325.1 XP_028690158.1 - 750 91
XM_028834326.1 XP_028690159.1 - 1043 91
XM_028834327.1 XP_028690160.1 - 725 91
GenBank mRNA JU320297.1 - - 721 91
JV046385.1 - - 716 91
JV046386.1 - - 730 91
JV497440.1 - - 767 91
JV636009.1 - - 671 91

View Full Product Details