Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESRRB
Entrez Gene ID
703799
Gene Name
estrogen related receptor beta
Gene Aliases
-
Chromosome Location
Chr.7: 138439820 - 138626537 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015144175.2 XP_014999661.1 - 875 143

View Full Product Details