Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM234A
Entrez Gene ID
694729
Gene Name
family with sequence similarity 234 member A
Gene Aliases
EGK_12279, ITFG3
Chromosome Location
Chr.20: 260781 - 293375 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015125147.2 XP_014980633.1 - 1353 92
XM_015125148.2 XP_014980634.1 - 1288 92
XM_015125150.2 XP_014980636.1 - 1358 92
XM_015125153.2 XP_014980639.1 - 1491 92
XM_015125154.2 XP_014980640.1 - 1418 92
XM_015125155.2 XP_014980641.1 - 1582 92
XM_028840357.1 XP_028696190.1 - 1483 92
XM_028840359.1 XP_028696192.1 - 1170 92
XM_028840360.1 XP_028696193.1 - 1353 92
GenBank mRNA JU472940.1 - - 1297 92
JV489103.1 - - 1167 92

View Full Product Details