Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPINK7
Entrez Gene ID
709674
Gene Name
serine peptidase inhibitor Kazal type 7
Gene Aliases
EGK_17000
Chromosome Location
Chr.6: 145842534 - 145844327 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015141206.2 XP_014996692.1 - 118 121

View Full Product Details