Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM83A
Entrez Gene ID
704023
Gene Name
family with sequence similarity 83 member A
Gene Aliases
-
Chromosome Location
Chr.8: 123493743 - 123527025 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194138.1 NP_001181067.1 - 1111 139
XM_028852219.1 XP_028708052.1 - 2641 139
XM_028852221.1 XP_028708054.1 - 1447 139

View Full Product Details