Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPK3BL2
Entrez Gene ID
715281
Gene Name
uroplakin 3B like 2
Gene Aliases
EGK_13574, UPK3BL, UPK3BL1
Chromosome Location
Chr.3: 44665723 - 44671878 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001108997.4 XP_001108997.2 - 952 103

View Full Product Details