Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HYAL1
Entrez Gene ID
702850
Gene Name
hyaluronidase 1
Gene Aliases
-
Chromosome Location
Chr.2: 106424033 - 106428067 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001102414.4 XP_001102414.2 - 1427 152

View Full Product Details