Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCNA
Entrez Gene ID
698393
Gene Name
germ cell nuclear acidic peptidase
Gene Aliases
ACRC
Chromosome Location
Chr.X: 68582656 - 68622432 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015127668.2 XP_014983154.1 - 2809 89

View Full Product Details