Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAB2
Entrez Gene ID
700197
Gene Name
GRB2 associated binding protein 2
Gene Aliases
-
Chromosome Location
Chr.14: 71295542 - 71498047 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015115335.2 XP_014970821.1 - 1665 148
GenBank mRNA JU497118.1 - - 1594 148
JU497119.1 - - 1594 148

View Full Product Details